(3-Chloro-2-fluorophenyl)acetic acid ≥97%
Supplier: Matrix Scientific
|
Warning
|
Synonyms:
2-(3-Chloro-2-fluorophenyl)acetic acid
002112-1G
002112-5G
002112-25G
101783-226EA
20
USD
101783-226
101783-228
101783-230
(3-Chloro-2-fluorophenyl)acetic acid ≥97%
(3-Chloro-2-fluorophenyl)acetic acid
Hazard code(s): Irritant
Formula:
(C₆H₃ClF)CH₂COOH MW: 188.59 g/mol |
MDL Number:
MFCD01631554 CAS Number: 261762-96-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...