1-Chloro-3-fluoro-4-iodobenzene ≥99%

Supplier: Matrix Scientific

Synonyms: 5-Chloro-1-fluoro-2-iodobenzene, 4-Chloro-2-fluoro-1-iodobenzene, 4-Chloro-2-fluoroiodobenzene

002109-25G 002109-100G
101783-212EA 44.62 USD
101783-212 101783-214
1-Chloro-3-fluoro-4-iodobenzene ≥99%
1-Chloro-3-fluoro-4-iodobenzene

Hazard code(s): Irritant

Formula: C₆H₃ClFI
MW: 256.45 g/mol
Boiling Pt: 107 °C (18mm)
Melting Pt: 17…19 °C
MDL Number: MFCD00079729
CAS Number: 6797-79-1

Order Now

Specification Test Results

Boiling point 107°C/18mm
Melting point 17-19°C
ND20 1.606

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR