4-Chloro-2-fluorobenzonitrile

Supplier: Matrix Scientific

002104-5G 002104-25G
101783-202EA 44.62 USD
101783-202 101783-204
4-Chloro-2-fluorobenzonitrile
4-Chloro-2-fluorobenzonitrile

Hazard code(s): Irritant

Formula: C₇H₃ClFN
MW: 155.56 g/mol
Boiling Pt: 273 °C (1013 hPa)
Melting Pt: 136 °C
MDL Number: MFCD00143284
CAS Number: 57381-51-8

Order Now

Specification Test Results

Boiling point 92°C/22mm
Melting point 59-61°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR