4-Chloro-2-fluorobenzaldehyde ≥97%

Supplier: Matrix Scientific
Warning

002101-25G 002101-100G
101783-198EA 60 USD
101783-198 101783-200
4-Chloro-2-fluorobenzaldehyde ≥97%
4-Chloro-2-fluorobenzaldehyde

Hazard code(s): Irritant, Air Sensitive

Formula: C₇H₄ClFO
MW: 158.56 g/mol
Melting Pt: 58…61 °C
MDL Number: MFCD00143282
CAS Number: 61072-56-8

Order Now

Specification Test Results

Melting point 58-61°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR