5-Chloro-2-fluorobenzonitrile ≥97%

Supplier: Matrix Scientific

002093-5G 002093-25G 002093-100G
101783-192EA 15.38 USD
101783-192 101783-194 102550-808
5-Chloro-2-fluorobenzonitrile ≥97%
5-Chloro-2-fluorobenzonitrile

Hazard code(s): Irritant

Formula: C₇H₃ClFN
Melting Pt: 67…70 °C
MDL Number: MFCD01631434
CAS Number: 57381-34-7
EINECS: 260-710-3
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR