3-Chloro-2-fluorobenzaldehyde ≥97%

Supplier: Matrix Scientific
Warning

002090-5G 002090-25G 002090-100G
101783-182EA 15.38 USD
101783-182 101783-184 101783-186
3-Chloro-2-fluorobenzaldehyde ≥97%
3-Chloro-2-fluorobenzaldehyde

Hazard code(s): Irritant, Air Sensitive

Formula: C₇H₄ClFO
MW: 158.56 g/mol
Density: 1.350 g/cm³
MDL Number: MFCD01631571
CAS Number: 85070-48-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR