5-Chloro-2-fluorobenzoic acid ≥97%

Supplier: Matrix Scientific
Warning

002088-5G 002088-25G 002088-100G
101783-176EA 18.46 USD
101783-176 101783-178 101783-180
5-Chloro-2-fluorobenzoic acid ≥97%
5-Chloro-2-fluorobenzoic acid

Hazard code(s): Irritant

Formula: C₇H₄ClFO₂
MDL Number: MFCD00665762
CAS Number: 394-30-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR