3-Chloro-2-fluorobenzoic acid ≥98%

Supplier: Matrix Scientific

002086-5G 002086-25G
101783-172EA 18.46 USD
101783-172 101783-174
3-Chloro-2-fluorobenzoic acid ≥98%
3-Chloro-2-fluorobenzoic acid

Hazard code(s): Irritant

Formula: C₇H₄ClFO₂
MW: 174.56 g/mol
Melting Pt: 176…178 °C
MDL Number: MFCD00042506
CAS Number: 161957-55-7

Order Now

Specification Test Results

Melting point 176-178°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR