3-Chloro-2-fluoro-6-(trifluoromethyl)benzoic acid

Supplier: Matrix Scientific

002074-1G 002074-5G
101783-160EA 76.92 USD
101783-160 101783-162
3-Chloro-2-fluoro-6-(trifluoromethyl)benzoic acid
3-Chloro-2-fluoro-6-(trifluoromethyl)benzoic acid
Formula: C₈H₃ClF₄O₂
MDL Number: MFCD01631342
CAS Number: 186517-41-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR