3-Chloro-2-fluoro-5-(trifluoromethyl)benzoic acid for synthesis

Supplier: Matrix Scientific

002063-1G 002063-5G
101783-136EA 52.31 USD
101783-136 101783-138
3-Chloro-2-fluoro-5-(trifluoromethyl)benzoic acid for synthesis
3-Chloro-2-fluoro-5-(trifluoromethyl)benzoic acid

Hazard code(s): Irritant

Formula: C₈H₃ClF₄O₂
Melting Pt: 89…91 °C
MDL Number: MFCD01631509
CAS Number: 129931-45-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR