6-Chloro-2-fluoro-m-toluic acid

Supplier: Matrix Scientific

Synonyms: 6-Chloro-2-fluoro-3-methylbenzoic acid

002051-1G 002051-5G
101783-112EA 30.77 USD
101783-112 101783-114
6-Chloro-2-fluoro-m-toluic acid
6-Chloro-2-fluoro-m-toluic acid
Formula: C₈H₆ClFO₂
MW: 188.59 g/mol
MDL Number: MFCD01631369
CAS Number: 32890-90-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR