2-Bromo-α,α,α,6-tetrafluorotoluene ≥97%

Supplier: Matrix Scientific

Synonyms: 1-Bromo-3-fluoro-2-(trifluoromethyl)benzene, 2-Bromo-6-fluorobenzotrifluoride

002014-5G 002014-25G
101783-060EA 52.31 USD
101783-060 101783-062
2-Bromo-α,α,α,6-tetrafluorotoluene ≥97%
2-Bromo-α,α,α,6-tetrafluorotoluene

Hazard code(s): Irritant

Formula: C₇H₃BrF₄
MW: 243 g/mol
MDL Number: MFCD01631569
CAS Number: 261951-85-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR