(5-Chloro-2-(trifluoromethyl)phenyl)acetic acid
Supplier: Matrix Scientific
|
Warning
|
002006-1G
002006-5G
101783-048EA
75.38
USD
101783-048
101783-050
(5-Chloro-2-(trifluoromethyl)phenyl)acetic acid
(5-Chloro-2-(trifluoromethyl)phenyl)acetic acid
Hazard code(s): Irritant
Formula:
C₉H₆ClF₃O₂ MW: 238.59 g/mol Melting Pt: 117…119 °C |
MDL Number:
MFCD01631358 CAS Number: 261763-25-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...