5-Chloro-2-(trifluoromethyl)benzoic acid ≥97% for synthesis

Supplier: Matrix Scientific

Synonyms: 5-Chloro-2-trifluoromethylbenzoic acid, 5-Chloro-α,α,α-trifluoro-o-toluic acid

002002-1G 002002-5G
101783-036EA 32.31 USD
101783-036 101783-038
5-Chloro-2-(trifluoromethyl)benzoic acid ≥97% for synthesis
5-Chloro-2-(trifluoromethyl)benzoic acid

Hazard code(s): Irritant

Formula: C₈H₄ClF₃O₂
MDL Number: MFCD01631353
CAS Number: 654-98-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR