(5-Fluoro-2-(trifluoromethyl)phenyl)acetic acid

Supplier: Matrix Scientific
Warning

Synonyms: 2-(5-Fluoro-2-(trifluoromethyl)phenyl)acetic acid

001999-1G 001999-5G
101783-028EA 47.69 USD
101783-028 101783-030
(5-Fluoro-2-(trifluoromethyl)phenyl)acetic acid
(5-Fluoro-2-(trifluoromethyl)phenyl)acetic acid
Formula: C₉H₆F₄O₂
MW: 222.14 g/mol
MDL Number: MFCD00236304
CAS Number: 239135-52-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR