5-Fluoro-2-(trifluoromethyl)benzyl alcohol ≥97% for synthesis

Supplier: Matrix Scientific

001994-5G 001994-25G
101783-016EA 56.92 USD
101783-016 101783-018
5-Fluoro-2-(trifluoromethyl)benzyl alcohol ≥97% for synthesis
5-Fluoro-2-(trifluoromethyl)benzyl alcohol

Hazard code(s): Irritant

Formula: C₈H₆F₄O
MDL Number: MFCD00236286
CAS Number: 238742-82-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR