α,α,α,5-Tetrafluoro-o-toluic acid ≥97%

Supplier: Matrix Scientific

001990-1G 001990-5G
101783-012EA 23.08 USD
101783-012 101783-014
α,α,α,5-Tetrafluoro-o-toluic acid ≥97%
α,α,α,5-Tetrafluoro-o-toluic acid

Hazard code(s): Irritant

Formula: C₈H₄F₄O₂
Melting Pt: 80…83 °C
MDL Number: MFCD00083521
CAS Number: 654-99-9

Order Now

Specification Test Results

Melting point 80-83°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR