(4-Fluoro-2-(trifluoromethyl)phenyl)acetic acid ≥98%

Supplier: Matrix Scientific
Warning

Synonyms: 2-(4-Fluoro-2-(trifluoromethyl)phenyl)acetic acid

001985-1G 001985-5G 001985-25G
101782-998EA 38.46 USD
101782-998 101783-000 101783-002
(4-Fluoro-2-(trifluoromethyl)phenyl)acetic acid ≥98%
(4-Fluoro-2-(trifluoromethyl)phenyl)acetic acid

Hazard code(s): Irritant

Formula: C₉H₆F₄O₂
MW: 222.14 g/mol
MDL Number: MFCD00061189
CAS Number: 195447-80-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR