4-Fluoro-2-(trifluoromethyl)benzaldehyde ≥97%
Supplier: Matrix Scientific
001979-5G
001979-25G
101782-982EA
44.62
USD
101782-982
101782-984
4-Fluoro-2-(trifluoromethyl)benzaldehyde ≥97%
4-Fluoro-2-(trifluoromethyl)benzaldehyde
Hazard code(s): Flammable, Air Sensitive
Formula:
C₈H₄F₄O Boiling Pt: 113…115 °C Density: 1.4 g/cm³ (20 °C) |
MDL Number:
MFCD00061313 CAS Number: 90176-80-0 UN: 1989 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...