α,α,α,4-Tetrafluoro-o-toluic acid ≥97%

Supplier: Matrix Scientific

001976-25G 001976-100G
101782-976EA 56.92 USD
101782-976 101782-978
α,α,α,4-Tetrafluoro-o-toluic acid ≥97%
α,α,α,4-Tetrafluoro-o-toluic acid

Hazard code(s): Irritant

Formula: C₈H₄F₄O₂
MW: 208.11 g/mol
Boiling Pt: 251 °C (1013 hPa)
Melting Pt: 121 - 124 °C
MDL Number: MFCD00040982
CAS Number: 141179-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR