3-Chloro-2,6-difluorophenol

Supplier: Matrix Scientific

001972-1G 001972-5G
101782-966EA 29.23 USD
101782-966 101782-968
3-Chloro-2,6-difluorophenol
3-Chloro-2,6-difluorophenol
Formula: C₆H₃ClF₂O
MW: 164.54 g/mol
MDL Number: MFCD01631330
CAS Number: 261762-51-0
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR