3-Chloro-2,6-difluorobenzoic acid ≥97%

Supplier: Matrix Scientific
Warning

001966-5G 001966-25G
101782-958EA 67.69 USD
101782-958 101782-960
3-Chloro-2,6-difluorobenzoic acid ≥97%
3-Chloro-2,6-difluorobenzoic acid

Hazard code(s): Irritant

Formula: C₇H₃ClF₂O₂
Melting Pt: 126…128 °C
MDL Number: MFCD01631322
CAS Number: 225104-76-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR