3-Chloro-2,6-dibromo-4-methylaniline ≥98%

Supplier: Matrix Scientific

Synonyms: 2,6-Dibromo-3-chloro-4-methylaniline, 3-Chloro-2,6-dibromo-p-toluidine, 1-Amino-3-chlor-2,6-dibrom-4-methylbenzene

001960-5G 001960-25G
101782-940EA 30.77 USD
101782-940 101782-942
3-Chloro-2,6-dibromo-4-methylaniline ≥98%
3-Chloro-2,6-dibromo-4-methylaniline

Hazard code(s): Irritant

Formula: C₇H₆Br₂ClN
Melting Pt: 76…78 °C
MDL Number: MFCD00051580
CAS Number: 84483-22-7

Order Now

Specification Test Results

Melting point 76-78°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR