3-Chloro-2,6-dibromo-4-fluoroaniline ≥97%

Supplier: Matrix Scientific

Synonyms: 2,6-Dibromo-3-chloro-4-fluoroaniline, 1-Amino-3-chlor-2,6-dibrom-4-fluorbenzol

001959-1G 001959-5G 001959-25G
101782-934EA 24.62 USD
101782-934 101782-936 101782-938
3-Chloro-2,6-dibromo-4-fluoroaniline ≥97%
3-Chloro-2,6-dibromo-4-fluoroaniline

Hazard code(s): Irritant

Formula: C₆H₃Br₂ClFN
MW: 303.36 g/mol
Boiling Pt: 302 °C (1013 hPa)
Melting Pt: 97 - 99 °C
MDL Number: MFCD00042199
CAS Number: 175135-09-8
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 97-99°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR