1-Chloro-2,5-dimethoxybenzene ≥98%
Supplier: Matrix Scientific
Synonyms:
2-Chloro-1,4-dimethoxybenzene
001958-25G
001958-100G
101782-930EA
36.92
USD
101782-930
101782-932
1-Chloro-2,5-dimethoxybenzene ≥98%
1-Chloro-2,5-dimethoxybenzene
Hazard code(s): Irritant
Formula:
C₈H₉ClO₂ MW: 172.61 g/mol Boiling Pt: 247 °C (1013 hPa) Melting Pt: 8.1 °C Density: 1.21 g/cm³ (20 °C) |
MDL Number:
MFCD00008356 CAS Number: 2100-42-7 EINECS: 218-267-9 |
Specification Test Results
Boiling point | 138°C/20mm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...