3-Acetyl-2,5-dichlorothiophene ≥98% for synthesis

Supplier: Matrix Scientific

Synonyms: 1-(2,5-Dichloro-3-thienyl)-1-ethanone

001952-25G 001952-100G
101782-920EA 36.92 USD
101782-920 101782-922
3-Acetyl-2,5-dichlorothiophene ≥98% for synthesis
3-Acetyl-2,5-dichlorothiophene

Hazard code(s): Irritant

Formula: C₆H₄Cl₂OS
Boiling Pt: 120…122 °C (6 mmHg)
Melting Pt: 37.5…39 °C
MDL Number: MFCD00014522
CAS Number: 36157-40-1
EINECS: 252-893-3

Order Now

Specification Test Results

Melting point 37.5-39°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR