4-Chloro-1,3-difluorobenzene ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Chloro-2,4-difluorobenzene

001950-25G 001950-100G
101782-914EA 21.54 USD
101782-914 101782-916
4-Chloro-1,3-difluorobenzene ≥95%
4-Chloro-1,3-difluorobenzene

Hazard code(s): Irritant

Formula: C₆H₃ClF₂
MW: 148.54 g/mol
Boiling Pt: 127 °C (1013 hPa)
Melting Pt: -26 °C
Density: 1.353 g/cm³ (20 °C)
MDL Number: MFCD00042569
CAS Number: 1435-44-5

Order Now

Specification Test Results

Boiling point 127-128°C
Density 1.353
ND20 1.475

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR