3-Chloro-2,4-difluorobenzoic acid ≥98%

Supplier: Matrix Scientific
Warning

001947-5G 001947-25G
101782-910EA 29.23 USD
101782-910 101782-912
3-Chloro-2,4-difluorobenzoic acid ≥98%
3-Chloro-2,4-difluorobenzoic acid

Hazard code(s): Irritant

Formula: C₇H₃ClF₂O₂
MW: 192.55 g/mol
Melting Pt: 174…176 °C
MDL Number: MFCD01631387
CAS Number: 154257-75-7

Order Now

Specification Test Results

Melting point 174-176°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR