1-Iodo-2,2,3,3-tetrafluoropropane ≥97%

Supplier: Matrix Scientific
Warning

001937-5G 001937-25G
101782-886EA 150.77 USD
101782-886 101782-888
1-Iodo-2,2,3,3-tetrafluoropropane ≥97%
1-Iodo-2,2,3,3-tetrafluoropropane

Hazard code(s): Irritant, Refrigerate

Formula: C₃H₃F₄I
MW: 241.95 g/mol
Boiling Pt: 95…97 °C
Density: 2.036 g/cm³
MDL Number: MFCD00019033
CAS Number: 679-87-8

Order Now

Specification Test Results

Boiling point 95-97°C
Density 2.036
ND20 1.412

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR