(±)-1-Phenyl-2,2,2-trifluoroethanol ≥98% for synthesis

Supplier: Matrix Scientific

Synonyms: α-(Trifluoromethyl)benzyl alcohol, 2,2,2-Trifluoro-1-phenylethan-1-ol

001933-5G 001933-25G
101782-878EA 115.38 USD
101782-878 101782-880
(±)-1-Phenyl-2,2,2-trifluoroethanol ≥98% for synthesis
(±)-1-Phenyl-2,2,2-trifluoroethanol

Hazard code(s): Irritant

Formula: C₆H₅CH(OH)CF₃
Boiling Pt: 57…59 °C (2mm)
Density: 1.293 g/cm³
MDL Number: MFCD00004498
CAS Number: 340-05-6
EINECS: 206-429-1

Order Now

Specification Test Results

Boiling point 64-65°C/5mm
Density 1.293
ND20 1.461


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR