4'-Bromo-2'-fluoroacetanilide ≥98%

Supplier: Matrix Scientific

Synonyms: N-(4-Bromo-2-fluorophenyl)acetamide

001930-25G 001930-100G
101782-868EA 38.46 USD
101782-868 101782-870
4'-Bromo-2'-fluoroacetanilide ≥98%
4'-Bromo-2'-fluoroacetanilide

Hazard code(s): Irritant

Formula: H₃CC(O)NH(C₆H₃BrF)
MW: 232.05 g/mol
Boiling Pt: 344 °C (1013 hPa)
Melting Pt: 158 - 160 °C
MDL Number: MFCD00042588
CAS Number: 326-66-9
EINECS: 206-314-6

Order Now

Specification Test Results

Melting point 142-146°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR