5-Fluoro-1-indanone ≥99% for synthesis

Supplier: Matrix Scientific

Synonyms: 5-Fluoro-2,3-dihydro-1H-inden-1-one

001918-5G 001918-25G 001918-100G
101782-848EA 58.46 USD
101782-848 101782-850 101782-852
5-Fluoro-1-indanone ≥99% for synthesis
5-Fluoro-1-indanone

Hazard code(s): Irritant, Refrigerate

Formula: C₉H₇FO
Boiling Pt: 113 - 114 °C (10 -rr)
Melting Pt: 35 - 37 °C
Density: 1.35 g/cm³ (20 °C)
MDL Number: MFCD00041031
CAS Number: 700-84-5

Order Now

Specification Test Results

Boiling point 93°C/3mm
Melting point 29-30°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR