5-Chloro-1-indanone ≥99%

Supplier: Matrix Scientific

001917-25G 001917-100G
101782-846EA 30.77 USD
101782-846 102550-804
5-Chloro-1-indanone ≥99%
5-Chloro-1-indanone

Hazard code(s): Irritant

Formula: C₉H₇ClO
MW: 166.61 g/mol
Boiling Pt: 150 °C (0.2 -rr)
Melting Pt: 100 °C
MDL Number: MFCD00041456
CAS Number: 42348-86-7

Order Now

Specification Test Results

Boiling point 125°C/3mm
Melting point 92-93°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR