1,2-Dibromo-1-chloro-1,2,2-trifluoroethane ≥99%
Supplier: Matrix Scientific
001903-25G
001903-100G
001903-500G
101782-832EA
43.08
USD
101782-832
101782-834
101782-836
1,2-Dibromo-1-chloro-1,2,2-trifluoroethane ≥99%
1,2-Dibromo-1-chloro-1,2,2-trifluoroethane
Hazard code(s): Irritant, Moisture Sensitive, Stored under Argon
Formula:
BrClFCCBrF₂ Boiling Pt: 93…94 °C Density: 2.2478 g/cm³ |
MDL Number:
MFCD00039316 CAS Number: 354-51-8 EINECS: 206-562-5 |
Specification Test Results
Boiling point | 93-94°C |
Density | 2.2478 |
ND20 | 1.4275 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...