1-Iodo-3,3,3-trifluoropropane ≥98%

Supplier: Matrix Scientific

001899-25G 001899-100G
101782-830EA 67.69 USD
101782-830 102663-546
1-Iodo-3,3,3-trifluoropropane ≥98%
1-Iodo-3,3,3-trifluoropropane

Hazard code(s): Irritant

Formula: C₃H₄F₃I
Boiling Pt: 90 °C
Density: 1.911 g/cm³
MDL Number: MFCD00038531
CAS Number: 460-37-7

Order Now

Specification Test Results

Boiling point 90°C
Density 1.911
ND20 1.422


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR