1,1,1-Trifluoroiodoethane ≥97%
Supplier: Matrix Scientific
|
Warning
|
Stabilization:
Stabilised with copper
001898-25G
001898-100G
101782-824EA
89.23
USD
101782-824
101782-826
1,1,1-Trifluoroiodoethane ≥97%
1,1,1-Trifluoroiodoethane
(Stabilized with copper)
Hazard code(s): Irritant
Formula:
C₂H₂F₃I Boiling Pt: 54.7 °C (1013 hPa) Density: 2.1499 g/cm³ (20 °C) |
MDL Number:
MFCD00001082 CAS Number: 353-83-3 EINECS: 206-541-0 UN: 2810 ADR: 6.1,III |
Specification Test Results
Boiling point | 55-56°C |
Density | 2.149 |
ND20 | 1.4001 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...