Dibenzo-30-crown-10 (dibenzo-1,4,7,10,13,16,19,22,25,28-decaoxacyclotriacontane) ≥98%

Supplier: Matrix Scientific

Synonyms: 6,7,9,10,12,13,15,16,23,24,26,27,29,30,32,33-Hexadecahydrodibenzo[b,q][1,4,7,10,13,16,19,22,25,28]decaoxacyclotriacontine, Dibenzo-30-crown-10-ether

001878-250MG 001878-1G
101782-792EA 124.62 USD
101782-792 101782-794
Dibenzo-30-crown-10 (dibenzo-1,4,7,10,13,16,19,22,25,28-decaoxacyclotriacontane) ≥98%
Dibenzo-30-crown-10 (dibenzo-1,4,7,10,13,16,19,22,25,28-decaoxacyclotriacontane)

Hazard code(s): Irritant

Formula: C₂₈H₄₀O₁₀
MW: 536.62 g/mol
Melting Pt: 106…108 °C
MDL Number: MFCD00054624
CAS Number: 17455-25-3

Order Now

Specification Test Results

Melting point 106-108°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR