2-Amino-2-thiazoline hydrochloride ≥99%

Supplier: Matrix Scientific

001869-25G 001869-100G
101782-774EA 83.08 USD
101782-774 101782-776
2-Amino-2-thiazoline hydrochloride ≥99%
2-Amino-2-thiazoline hydrochloride

Hazard code(s): Irritant

Formula: C₃H₆N₂S
Melting Pt: 204…205 °C
MDL Number: MFCD00012709
CAS Number: 3882-98-2
EINECS: 217-224-1

Order Now

Specification Test Results

Melting point 204-205°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR