4-tert-Butyl-1-bromobenzene ≥98%
Supplier: Matrix Scientific
Synonyms:
1-Bromo-4-tert-butylbenzene, 2-(4-Bromophenyl)-2-methylpropane, 1-Bromo-4-(tert-butyl)benzene, 4-Bromo-tert-butylbenzene
001868-25G
001868-100G
101782-770EA
24.62
USD
101782-770
101782-772
4-tert-Butyl-1-bromobenzene ≥98%
4-tert-Butyl-1-bromobenzene
Hazard code(s): Irritant
Formula:
C₁₀H₁₃Br MW: 213.12 g/mol Boiling Pt: 232 °C (1013 hPa) Melting Pt: 15.5 °C Density: 1.2572 g/cm³ (15 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000108 CAS Number: 3972-65-4 EINECS: 223-599-2 |
Specification Test Results
Boiling point | 80-81°C/12m |
Melting point | 15-16°C |
Density | 1.229 |
ND20 | 1.533 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...