4-Nitro-2-picoline-N-oxide ≥98% for synthesis

Supplier: Matrix Scientific

Synonyms: 2-Methyl-4-nitropyridine-1-oxide, 4-Nitro-2-picoline N-oxide, 2-Methyl-4-nitropyridine-N-oxide

001862-5G 001862-25G 001862-100G
101782-760EA 30.77 USD
101782-760 101782-762 101782-764
4-Nitro-2-picoline-N-oxide ≥98% for synthesis
4-Nitro-2-picoline-N-oxide

Hazard code(s): Irritant, Stored under Argon

Formula: C₆H₆N₂O₃
Melting Pt: 156 °C
MDL Number: MFCD00014625
CAS Number: 5470-66-6

Order Now

Specification Test Results

Melting point 154-156°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR