4-Nitro-3-picoline-N-oxide ≥98% for synthesis
Supplier: Matrix Scientific
Synonyms:
4-Nitropicoline N-oxide, 3-Methyl-4-nitropyridine 1-oxide, 3-Methyl-4-nitropyridine-N-oxide, 3-Methyl-4-nitropyridine N-oxide, 4-Nitro-3-picoline N-oxide, 3-Methyl-4-nitropyridine-1-oxide
001861-5G
001861-25G
101782-756EA
21.54
USD
101782-756
101782-758
4-Nitro-3-picoline-N-oxide ≥98% for synthesis
4-Nitro-3-picoline-N-oxide
Hazard code(s): Toxic
Formula:
C₆H₆N₂O₃ MW: 154.13 g/mol Melting Pt: 137.5 |
MDL Number:
MFCD00014626 CAS Number: 1074-98-2 EINECS: 214-050-8 UN: 2811 ADR: 6.1 III |
Specification Test Results
Melting point | 132-135°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...