2-Amino-4-phenylthiazole ≥97%

Supplier: Matrix Scientific

001859-5G 001859-25G
101782-752EA 89.23 USD
101782-752 101782-754
2-Amino-4-phenylthiazole ≥97%
2-Amino-4-phenylthiazole

Hazard code(s): Toxic

Formula: C₉H₈N₂S
Melting Pt: 149…152 °C
MDL Number: MFCD00039680
CAS Number: 2010-06-2
EINECS: 217-926-8
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 149-152°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR