4-Bromo-3-nitrotoluene ≥99% for synthesis
Supplier: Matrix Scientific
|
Warning
|
Synonyms:
1-Bromo-4-methyl-2-nitrobenzene
001855-25G
001855-100G
101782-740EA
29.23
USD
101782-740
101782-742
4-Bromo-3-nitrotoluene ≥99% for synthesis
4-Bromo-3-nitrotoluene
Hazard code(s): Irritant
Formula:
C₇H₆BrNO₂ MW: 216.03 g/mol Boiling Pt: 96 °C (1mm) Melting Pt: 35 Density: 1.57 g/cm³ (20 °C) |
MDL Number:
MFCD00024180 CAS Number: 5326-34-1 EINECS: 226-203-6 |
Specification Test Results
Boiling point | 96°C/1mm |
Melting point | 30-32°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...