2-Bromo-4-nitrotoluene ≥98% for synthesis
Supplier: Matrix Scientific
|
Warning
|
001854-25G
001854-100G
101782-736EA
23.08
USD
101782-736
101782-738
2-Bromo-4-nitrotoluene ≥98% for synthesis
2-Bromo-4-nitrotoluene
Hazard code(s): Irritant
Formula:
C₇H₆BrNO₂ Boiling Pt: 175 °C (38 -rr) Melting Pt: 77.5 °C |
MDL Number:
MFCD00007195 CAS Number: 7745-93-9 EINECS: 231-809-9 |
Specification Test Results
Melting point | 76-77°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...