2-Bromo-5-nitrotoluene ≥98% for synthesis

Supplier: Matrix Scientific

Synonyms: 1-Bromo-2-methyl-4-nitrobenzene

001853-25G 001853-100G
101782-732EA 60 USD
101782-732 101782-734
2-Bromo-5-nitrotoluene ≥98% for synthesis
2-Bromo-5-nitrotoluene

Hazard code(s): Irritant

Formula: C₇H₆BrNO₂
Boiling Pt: 140 - 143 °C (17 -rr)
Melting Pt: 78 °C
MDL Number: MFCD00007281
CAS Number: 7149-70-4
EINECS: 230-481-4

Order Now

Specification Test Results

Melting point 78-80°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR