5-Bromo-α,α,α-trifluoro-2-nitrotoluene ≥98%

Supplier: Matrix Scientific

Synonyms: 1-Bromo-4-nitro-3-(trifluoromethyl)benzene, 5-Bromo-2-nitrobenzotrifluoride

001851-25G 001851-100G
101782-728EA 26.15 USD
101782-728 101782-730
5-Bromo-α,α,α-trifluoro-2-nitrotoluene ≥98%
5-Bromo-α,α,α-trifluoro-2-nitrotoluene

Hazard code(s): Irritant

Formula: C₇H₃BrF₃NO₂
MW: 270.01 g/mol
Boiling Pt: 95…100 °C (5mm)
Melting Pt: 33…35 °C
Density: 1.799 g/cm³ (25 °C)
MDL Number: MFCD00014706
CAS Number: 344-38-7

Order Now

Specification Test Results

Boiling point 95-100°C/5m
Melting point 33-35°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR