2-Nitro-4-(trifluoromethyl)aniline ≥97%
Supplier: Matrix Scientific
001847-25G
001847-100G
101782-718EA
32.31
USD
101782-718
102832-742
2-Nitro-4-(trifluoromethyl)aniline ≥97%
2-Nitro-4-(trifluoromethyl)aniline
Hazard code(s): Toxic
Formula:
C₇H₅F₃N₂O₂ Melting Pt: 107 °C |
MDL Number:
MFCD00007155 CAS Number: 400-98-6 EINECS: 206-926-3 UN: 2811 ADR: 6.1,III |
Specification Test Results
Melting point | 105-106°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...