4-Bromo-3-nitrobenzotrifluoride ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Bromo-3-nitrobenzotrifluoride, 4-Bromo-α,α,α-trifluoro-3-nitrotoluene, 1-Bromo-2-nitro-4-(trifluoromethyl)benzene

001844-25G 001844-100G
101782-708EA 29.23 USD
101782-708 101782-710
4-Bromo-3-nitrobenzotrifluoride ≥97%
4-Bromo-3-nitrobenzotrifluoride

Hazard code(s): Irritant

Formula: C₇H₃BrF₃NO₂
MW: 270.01 g/mol
Boiling Pt: 102…105 °C (10 mmHg)
Density: 1.76 g/cm³ (20 °C)
MDL Number: MFCD00014685
CAS Number: 349-03-1

Order Now

Specification Test Results

Density 1.763
ND20 1.511

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR