4-Bromo-2-nitroanisole ≥98%

Supplier: Matrix Scientific

Synonyms: 4-Bromo-1-methoxy-2-nitrobenzene

001843-5G 001843-25G
101782-704EA 16.92 USD
101782-704 101782-706
4-Bromo-2-nitroanisole ≥98%
4-Bromo-2-nitroanisole

Hazard code(s): Irritant

Formula: C₇H₆BrNO₃
MW: 232.03 g/mol
Melting Pt: 87…88 °C
MDL Number: MFCD00055529
CAS Number: 33696-00-3
EINECS: 251-642-5

Order Now

Specification Test Results

Melting point 87-88°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR