2-Bromo-4-nitroanisole ≥98%

Supplier: Matrix Scientific

Synonyms: 2-Bromo-1-methoxy-4-nitrobenzene

001842-25G 001842-100G
101782-700EA 29.23 USD
101782-700 101782-702
2-Bromo-4-nitroanisole ≥98%
2-Bromo-4-nitroanisole

Hazard code(s): Irritant

Formula: C₇H₆BrNO₃
MW: 232.03 g/mol
Melting Pt: 105…108 °C
MDL Number: MFCD00041242
CAS Number: 5197-28-4
EINECS: 225-983-5

Order Now

Specification Test Results

Melting point 105-108°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR